Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562405.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000562574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000565053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008854.hg.1 |
ENST00000569249.1 |
Ensembl |