Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031379.hg.1 |
ENST00000262189.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031379.hg.1 |
ENST00000355193.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031379.hg.1 |
NM_170606.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07031379.hg.1 |
TC07002045.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000262189.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000355193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000473186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000558084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
NM_170606.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
TC0700013102.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000558084.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000262189.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000355193.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
ENST00000473186.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700194590.hg.1 |
NM_170606.2 |
Refseq |