Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01005535_m1
          See other KMT2C GE Assays ›
          Gene Symbol:
          KMT2C
          Gene Name:
          lysine methyltransferase 2C
          Gene Aliases:
          HALR, MLL3
          Chromosome Location:
          Chr.7: 152134925 - 152436642 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01005535_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KMT2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170606.2 NP_733751.2 51 - 52 12989 132
          XM_005250025.4 XP_005250082.1 54 - 55 13008 132
          XM_005250026.3 XP_005250083.1 54 - 55 13042 132
          XM_005250027.4 XP_005250084.1 53 - 54 13213 132
          XM_005250028.4 XP_005250085.1 53 - 54 13216 132
          XM_005250031.4 XP_005250088.1 52 - 53 13051 132
          XM_006716077.3 XP_006716140.1 53 - 54 13213 132
          XM_006716078.3 XP_006716141.1 53 - 54 13144 132
          XM_006716079.3 XP_006716142.1 52 - 53 13048 132
          XM_011516450.2 XP_011514752.1 52 - 53 13168 132
          XM_011516451.2 XP_011514753.1 52 - 53 13096 132
          XM_011516452.2 XP_011514754.1 51 - 52 13063 132
          XM_011516453.2 XP_011514755.1 52 - 53 12979 132
          XM_011516454.2 XP_011514756.1 48 - 49 12211 132
          XM_011516456.2 XP_011514758.1 52 - 53 13168 132
          XM_017012480.1 XP_016867969.1 54 - 55 13045 132
          XM_017012481.1 XP_016867970.1 54 - 55 13005 132
          XM_017012482.1 XP_016867971.1 53 - 54 13213 132
          XM_017012483.1 XP_016867972.1 53 - 54 13213 132
          XM_017012484.1 XP_016867973.1 53 - 54 13183 132
          XM_017012485.1 XP_016867974.1 52 - 53 13165 132
          XM_017012486.1 XP_016867975.1 53 - 54 13141 132
          XM_017012487.1 XP_016867976.1 53 - 54 13184 132
          XM_017012488.1 XP_016867977.1 52 - 53 13033 132
          XM_017012489.1 XP_016867978.1 34 - 35 9723 132
          XM_017012490.1 XP_016867979.1 31 - 32 9295 132
          GenBank mRNA AF264750.2 - 38 - 39 10487 132
          AL832387.1 - 15 - 16 5452 132
          AL833924.1 - 8 - 9 1136 132
          AY024361.1 - 51 - 52 12950 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013102.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07002045.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KMT2C

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 methylation
          DNA binding
          RNA binding
          protein binding
          zinc ion binding
          acyltransferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H3-K4 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07031353.hg.1 ENST00000262189.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031353.hg.1 ENST00000355193.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031353.hg.1 NM_170606.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031353.hg.1 TC07002045.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000262189.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000355193.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000360104.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000424877.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000473186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000558084.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 NM_170606.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 TC0700013102.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000355193.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000262189.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000360104.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000424877.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000473186.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 ENST00000558084.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194558.hg.1 NM_170606.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0