Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032260.hg.1 |
ENST00000356763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032260.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032260.hg.1 |
TC03001778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032262.hg.1 |
ENST00000356763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032262.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032262.hg.1 |
TC03001778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032263.hg.1 |
ENST00000356763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032263.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032263.hg.1 |
TC03001778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032261.hg.1 |
ENST00000356763.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032261.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03032261.hg.1 |
TC03001778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000504834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000504834.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192958.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192958.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192958.hg.1 |
TC0300012426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192958.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192958.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192955.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192955.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192955.hg.1 |
TC0300012426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192955.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192955.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000504834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000504834.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000507330.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000508273.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000511760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512362.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
ENST00000512677.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012426.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192957.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192957.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192957.hg.1 |
TC0300012426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192957.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192957.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192956.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192956.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192956.hg.1 |
TC0300012426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192956.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192956.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192959.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192959.hg.1 |
NM_014602.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300192959.hg.1 |
TC0300012426.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300192959.hg.1 |
ENST00000356763.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300192959.hg.1 |
NM_014602.2 |
Refseq |