Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01013460_s1
          See other FOXC2 GE Assays ›
          Gene Symbol:
          FOXC2
          Gene Name:
          forkhead box C2
          Gene Aliases:
          FKHL14, LD, MFH-1, MFH1
          Chromosome Location:
          Chr.16: 86567251 - 86568933 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01013460_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FOXC2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005251.2 NP_005242.1 1 170 85
          GenBank mRNA BC113437.1 - 1 170 8322489 85
          BC113439.1 - 1 170 8322491 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600008738.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000670.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FOXC2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          ossification
          branching involved in blood vessel morphogenesis
          metanephros development
          ureteric bud development
          somitogenesis
          lymphangiogenesis
          blood vessel remodeling
          transcription by RNA polymerase II
          Notch signaling pathway
          mesoderm development
          heart development
          insulin receptor signaling pathway
          response to hormone
          positive regulation of endothelial cell migration
          neural crest cell development
          collagen fibril organization
          positive regulation of cell adhesion mediated by integrin
          embryonic heart tube development
          positive regulation of vascular wound healing
          camera-type eye development
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          regulation of organ growth
          vascular endothelial growth factor receptor signaling pathway
          paraxial mesodermal cell fate commitment
          embryonic viscerocranium morphogenesis
          artery morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle cell proliferation
          glomerular endothelium development
          glomerular visceral epithelial cell differentiation
          glomerular mesangial cell development
          positive regulation of cell migration involved in sprouting angiogenesis
          blood vessel diameter maintenance
          negative regulation of apoptotic process involved in outflow tract morphogenesis
          transcription cis-regulatory region binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA-binding transcription factor activity
          protein binding
          chromatin DNA binding
          identical protein binding
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009458.hg.1 ENST00000320354.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16009458.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009458.hg.1 TC16000670.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16009457.hg.1 ENST00000320354.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16009457.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009457.hg.1 TC16000670.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16009459.hg.1 ENST00000320354.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16009459.hg.1 NM_005251.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16009459.hg.1 TC16000670.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay TC1600008738.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600165244.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165244.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600165244.hg.1 TC1600008738.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600165244.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165244.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600165246.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165246.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600165246.hg.1 TC1600008738.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600165246.hg.1 ENST00000320354.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1600165246.hg.1 NM_005251.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 ENST00000380874.3 Ensembl
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq
          Affymetrix Human Clariom D Assay TC0600006554.hg.1 NM_001453.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0