Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034501.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034501.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034501.hg.1 |
TC03001920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034502.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034502.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034502.hg.1 |
TC03001920.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034503.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034503.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03034503.hg.1 |
TC03001920.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0300196187.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196187.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196187.hg.1 |
TC0300012860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196187.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196187.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196188.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196188.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196188.hg.1 |
TC0300012860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196188.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196188.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196189.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196189.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300196189.hg.1 |
TC0300012860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300196189.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300196189.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
ENST00000389740.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012860.hg.1 |
NM_001038705.2 |
Refseq |