Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01026995_g1
          See other JAK1 GE Assays ›
          Gene Symbol:
          JAK1
          Gene Name:
          Janus kinase 1
          Gene Aliases:
          JAK1A, JAK1B, JTK3
          Chromosome Location:
          Chr.1: 64833223 - 65067746 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01026995_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: JAK1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001320923.1 NP_001307852.1 24 - 25 3471 72
          NM_001321852.1 NP_001308781.1 23 - 24 3474 72
          NM_001321853.1 NP_001308782.1 25 - 26 3733 72
          NM_001321854.1 NP_001308783.1 24 - 25 3649 72
          NM_001321855.1 NP_001308784.1 24 - 25 3652 72
          NM_001321856.1 NP_001308785.1 23 - 24 3387 72
          NM_001321857.1 NP_001308786.1 23 - 24 3565 72
          NM_002227.3 NP_002218.2 23 - 24 3568 72
          XM_017001212.1 XP_016856701.1 23 - 24 3412 72
          GenBank mRNA AB209057.1 - 22 - 23 3324 72
          AB219242.1 - 21 - 22 3215 72
          AK314257.1 - 24 - 25 3472 72
          BC132729.1 - 22 - 23 3286 40146751 72
          BX647675.1 - 9 - 10 1172 72
          HQ628631.1 - 21 - 22 3248 72
          M64174.1 - 22 - 23 3293 72

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene JAK1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell migration
          cell differentiation
          intracellular signal transduction
          peptidyl-tyrosine autophosphorylation
          interleukin-2-mediated signaling pathway
          regulation of cell population proliferation
          positive regulation of GTPase activity
          innate immune response
          response to antibiotic
          interferon-gamma-mediated signaling pathway
          regulation of interferon-gamma-mediated signaling pathway
          type I interferon signaling pathway
          regulation of type I interferon-mediated signaling pathway
          positive regulation of sprouting angiogenesis
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signaling receptor binding
          growth hormone receptor binding
          protein binding
          ATP binding
          protein phosphatase binding
          ubiquitin protein ligase binding
          CCR5 chemokine receptor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000467764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000467764.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000481702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000481702.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000467764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000467764.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000471473.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000481702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000481702.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000494904.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 NM_002227.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000342505.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100014457.hg.1 ENST00000465376.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0