Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01028185_m1
          See other NRXN3 GE Assays ›
          Gene Symbol:
          NRXN3
          Gene Name:
          neurexin 3
          Gene Aliases:
          C14orf60
          Chromosome Location:
          Chr.14: 77979904 - 79868290 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01028185_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NRXN3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004796.5 NP_004787.2 12 - 13 2656 91
          NR_073547.1 14 - 15 4237 91
          XM_005268218.2 XP_005268275.1 17 - 18 4306 91
          XM_006720322.2 XP_006720385.1 16 - 17 4294 91
          XM_006720323.2 XP_006720386.1 15 - 16 4270 91
          XM_011537363.1 XP_011535665.1 17 - 18 4306 91
          XM_011537364.2 XP_011535666.1 18 - 19 4268 91
          XM_011537365.2 XP_011535667.1 17 - 18 4094 91
          XM_011537366.1 XP_011535668.1 16 - 17 4282 91
          XM_011537367.1 XP_011535669.1 17 - 18 4306 91
          XM_011537368.1 XP_011535670.1 17 - 18 4306 91
          XM_011537369.1 XP_011535671.1 17 - 18 4306 91
          XM_011537370.1 XP_011535672.1 16 - 17 4282 91
          XM_011537371.1 XP_011535673.1 17 - 18 4306 91
          XM_011537372.1 XP_011535674.1 17 - 18 4306 91
          XM_011537373.1 XP_011535675.1 17 - 18 4306 91
          XM_011537377.2 XP_011535679.1 13 - 14 2598 91
          XM_017021790.1 XP_016877279.1 18 - 19 4406 91
          XM_017021791.1 XP_016877280.1 18 - 19 4189 91
          XM_017021792.1 XP_016877281.1 16 - 17 4089 91
          XM_017021793.1 XP_016877282.1 16 - 17 4279 91
          XM_017021794.1 XP_016877283.1 15 - 16 4270 91
          XM_017021795.1 XP_016877284.1 15 - 16 4065 91
          XM_017021796.1 XP_016877285.1 15 - 16 4255 91
          XM_017021797.1 XP_016877286.1 14 - 15 4246 91
          XM_017021798.1 XP_016877287.1 14 - 15 4243 91
          XM_017021799.1 XP_016877288.1 17 - 18 4306 91
          XM_017021800.1 XP_016877289.1 17 - 18 4306 91
          XM_017021801.1 XP_016877290.1 16 - 17 4279 91
          XM_017021802.1 XP_016877291.1 15 - 16 4300 91
          XM_017021803.1 XP_016877292.1 16 - 17 4285 91
          XM_017021804.1 XP_016877293.1 16 - 17 4279 91
          XM_017021805.1 XP_016877294.1 15 - 16 4270 91
          XM_017021806.1 XP_016877295.1 15 - 16 4255 91
          XM_017021807.1 XP_016877296.1 17 - 18 4306 91
          GenBank mRNA AB018286.1 - 12 - 13 2637 91
          AJ316284.2 - 14 - 15 4025 91
          BC152457.1 - 12 - 13 2637 8862590 91

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene NRXN3

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron cell-cell adhesion
          neurotransmitter secretion
          axon guidance
          synapse assembly
          learning
          adult behavior
          social behavior
          positive regulation of synapse assembly
          vocalization behavior
          positive regulation of synapse maturation
          calcium channel regulator activity
          signaling receptor activity
          metal ion binding
          cell adhesion molecule binding
          neuroligin family protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix