Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
223911_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006487.hg.1 |
ENST00000624747.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006487.hg.1 |
ENST00000624747.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000567091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008970.hg.1 |
ENST00000571933.1 |
Ensembl |
Human Genome U133 Plus 2.0 Array |
223911_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
223911_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
223911_at6 |
- |
- |
Human Genome U95 Set |
57469_at |
- |
- |
Human Genome U95 Set |
57469_PM_at |
- |
- |
Human Genome U133 Set |
223911_at |
- |
- |
Human Genome U133 Set |
223911_PM_at |
- |
- |