Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000287898.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000296794.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000296799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000426542.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000437974.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000512883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000513042.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
ENST00000545377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
NM_001080479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
NM_001177693.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
NM_001244364.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05004903.hg.1 |
TC05000355.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000296794.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000296799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000426542.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000437974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000510312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000512883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000513042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000545377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001080479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001177693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001244364.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
TC0500007780.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000437974.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000296794.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000296799.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000426542.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000510312.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000512883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000513042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
ENST00000545377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001080479.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001177693.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500153129.hg.1 |
NM_001244364.1 |
Refseq |