Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8076814 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8076813 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1561099_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009033.hg.1 |
ENST00000625149.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800011073.hg.1 |
ENST00000517884.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165436.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165436.hg.1 |
TC2200009033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165436.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165438.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165438.hg.1 |
TC2200009033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165438.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009033.hg.1 |
ENST00000625149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009033.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165437.hg.1 |
ENST00000625149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165437.hg.1 |
NR_122047.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200165437.hg.1 |
TC2200009033.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200165437.hg.1 |
ENST00000625149.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200165437.hg.1 |
NR_122047.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
1561099_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1561099_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1561099_at4 |
- |
- |