Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
ENST00000320476.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
ENST00000356994.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
ENST00000377533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
ENST00000377539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
NM_015356.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
NM_182706.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08022829.hg.1 |
TC08001732.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000320476.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000356994.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000377533.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000526832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
NM_015356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
NM_182706.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
TC0800012165.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000320476.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000356994.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000377533.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
ENST00000526832.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
NM_015356.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800183022.hg.1 |
NM_182706.4 |
Refseq |