Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01036300_g1
          See other ETS2 GE Assays ›
          Gene Symbol:
          ETS2
          Gene Name:
          ETS proto-oncogene 2, transcription factor
          Gene Aliases:
          ETS2IT1
          Chromosome Location:
          Chr.21: 38805307 - 38824955 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01036300_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ETS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256295.1 NP_001243224.1 4 - 5 632 85
          NM_005239.5 NP_005230.1 3 - 4 462 85
          XM_005260935.1 XP_005260992.1 3 - 4 535 85
          XM_017028290.1 XP_016883779.1 3 - 4 3132 85
          GenBank mRNA AK096841.1 - 4 - 5 632 85
          AK315563.1 - 3 - 4 365 85
          BC017040.1 - 3 - 4 334 3852274 85
          BC042954.2 - 3 - 4 364 5276704 85
          BT006838.1 - 2 - 3 172 85
          J04102.1 - 3 - 4 463 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007140.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000169.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ETS2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          skeletal system development
          ectodermal cell fate commitment
          transcription, DNA-templated
          mesoderm development
          cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          primitive streak formation
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          glucocorticoid receptor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001614.hg.1 TC21000169.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21001614.hg.1 ENST00000360214.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001614.hg.1 ENST00000360938.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001614.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR21001614.hg.1 NM_005239.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010905.hg.1 ENST00000439300.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010905.hg.1 ENST00000439300.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 TC2100007140.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100148160.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC0X00010905.hg.1 ENST00000439300.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010905.hg.1 ENST00000439300.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360214.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000360938.7 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000432278.5 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 ENST00000456966.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_001256295.1 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Affymetrix Human Clariom D Assay TC2100007140.hg.1 NM_005239.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 161027 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix