Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006092.2 | NP_006083.1 | 8 - 9 | 2889 | 83 | |
XM_005249568.1 | XP_005249625.1 | 9 - 10 | 2944 | 83 | ||
XM_005249572.1 | XP_005249629.1 | 7 - 8 | 2807 | 83 | ||
XM_005249576.1 | XP_005249633.1 | 4 - 5 | 1706 | 83 | ||
XM_006715633.2 | XP_006715696.1 | 9 - 10 | 2981 | 83 | ||
XM_011515079.1 | XP_011513381.1 | 10 - 11 | 3127 | 83 | ||
XM_011515080.2 | XP_011513382.1 | 8 - 9 | 2803 | 83 | ||
XM_011515081.2 | XP_011513383.1 | 8 - 9 | 2953 | 83 | ||
XM_011515084.1 | XP_011513386.1 | 9 - 10 | 2944 | 83 | ||
XM_011515085.1 | XP_011513387.1 | 9 - 10 | 2944 | 83 | ||
XM_011515087.1 | XP_011513389.1 | 9 - 10 | 2944 | 83 | ||
XM_017011674.1 | XP_016867163.1 | 9 - 10 | 2944 | 83 | ||
XM_017011675.1 | XP_016867164.1 | 8 - 9 | 2850 | 83 | ||
GenBank mRNA | AF113925.1 | - | 8 - 9 | 2790 | 83 | |
AF126484.1 | - | 8 - 9 | 2817 | 83 | ||
BC040339.1 | - | 8 - 9 | 2804 | 4338930 | 83 | |
KU178514.1 | - | 5 - 6 | 2366 | 83 | ||
KU178515.1 | - | 4 - 5 | 541 | 83 | ||
KU178517.1 | - | 4 - 5 | 541 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |