Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01037069_m1
          See other NR1H4 GE Assays ›
          Gene Symbol:
          NR1H4
          Gene Name:
          nuclear receptor subfamily 1 group H member 4
          Gene Aliases:
          BAR, FXR, HRR-1, HRR1, RIP14
          Chromosome Location:
          Chr.12: 100473773 - 100563867 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01037069_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NR1H4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001206977.1 NP_001193906.1 6 - 7 1237 112
          NM_001206979.1 NP_001193908.1 5 - 6 1070 112
          NM_001206993.1 NP_001193922.1 3 - 4 652 112
          XM_006719719.2 XP_006719782.1 3 - 4 653 112
          XM_011539040.2 XP_011537342.1 5 - 6 1988 112
          GenBank mRNA AF384555.1 - 3 - 4 594 112
          AF478446.1 - 3 - 4 652 112
          AK296612.1 - 3 - 4 653 112
          AK296647.1 - 3 - 4 588 112
          BC071778.1 - 3 - 4 592 112
          BC130573.1 - 5 - 6 927 40146604 112
          HQ709175.1 - 3 - 4 594 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200008595.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000778.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          nitrogen catabolite activation of transcription from RNA polymerase II promoter
          regulation of carbohydrate metabolic process
          transcription initiation from RNA polymerase II promoter
          inflammatory response
          signal transduction
          Notch signaling pathway
          bile acid metabolic process
          response to glucose
          regulation of low-density lipoprotein particle clearance
          bile acid and bile salt transport
          intracellular receptor signaling pathway
          response to lipopolysaccharide
          interleukin-17 production
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          toll-like receptor 4 signaling pathway
          regulation of urea metabolic process
          histone H3-R17 methylation
          cellular triglyceride homeostasis
          bile acid signaling pathway
          intracellular bile acid receptor signaling pathway
          negative regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          steroid hormone mediated signaling pathway
          innate immune response
          positive regulation of transcription by RNA polymerase II
          digestive tract development
          regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of bile acid biosynthetic process
          negative regulation of bile acid biosynthetic process
          cellular response to fatty acid
          cellular response to organonitrogen compound
          regulation of cholesterol metabolic process
          positive regulation of glutamate metabolic process
          positive regulation of ammonia assimilation cycle
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA-binding transcription factor activity
          steroid hormone receptor activity
          transcription coactivator activity
          transcription corepressor activity
          nuclear receptor activity
          protein binding
          zinc ion binding
          nuclear receptor binding
          bile acid binding
          bile acid receptor activity
          peptide binding
          sequence-specific DNA binding
          retinoid X receptor binding
          chenodeoxycholic acid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 ENST00000188403.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 ENST00000392986.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 ENST00000548884.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 ENST00000549996.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 ENST00000551379.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_001206977.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_001206978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_001206979.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_001206992.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_001206993.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 NM_005123.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12009793.hg.1 TC12000778.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000188403.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000321046.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000392986.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000548884.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000549996.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000551379.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206978.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206979.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206992.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206993.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_005123.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 TC1200008595.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000548884.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000188403.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000321046.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000392986.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000549996.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 ENST00000551379.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206977.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206978.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206979.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206992.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_001206993.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200163210.hg.1 NM_005123.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0