Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01046445_m1
          See other ARMC8 GE Assays ›
          Gene Symbol:
          ARMC8
          Gene Name:
          armadillo repeat containing 8
          Gene Aliases:
          GID5, HSPC056, S863-2, VID28
          Chromosome Location:
          Chr.3: 138187248 - 138298389 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01046445_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARMC8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001267041.2 NP_001253970.1 2 - 3 427 77
          NM_001267042.2 NP_001253971.1 2 - 3 427 77
          NM_014154.4 NP_054873.2 3 - 4 503 77
          NM_015396.5 NP_056211.2 3 - 4 503 77
          NM_213654.3 NP_998819.1 3 - 4 557 77
          XM_006713564.3 XP_006713627.1 2 - 3 422 77
          XM_006713565.2 XP_006713628.1 3 - 4 463 77
          XM_006713567.3 XP_006713630.1 2 - 3 422 77
          XM_017006095.1 XP_016861584.1 3 - 4 425 77
          XM_017006100.1 XP_016861589.1 2 - 3 422 77
          XM_017006101.1 XP_016861590.1 3 - 4 425 77
          GenBank mRNA AF109184.1 - 3 - 4 256 77
          AF161541.1 - 3 - 4 222 77
          AK289577.1 - 3 - 4 199 77
          AK296757.1 - 2 - 3 158 77
          AK296837.1 - 2 - 3 127 77
          BC013424.1 - 3 - 4 257 3892143 77
          BC032661.1 - 3 - 4 367 5530425 77
          BC041699.1 - 3 - 4 182 5240423 77
          BF966252.1 - 2 - 3 380 77
          CX167429.1 - 2 - 3 132 77
          DC304503.1 - 3 - 4 480 77
          DC342329.1 - 3 - 4 468 77
          HY004298.1 - 2 - 3 428 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as HTA transcript cluster(s) TC03000743.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ARMC8

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000358441.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000393058.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000461822.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000469044.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000470821.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000471453.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000481646.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000485396.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000489213.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000491704.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 ENST00000538260.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 NM_014154.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 NM_015396.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 NM_213654.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03013459.hg.1 TC03000743.hg.1 HTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0