Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01051875_m1
          See other ITSN1 GE Assays ›
          Gene Symbol:
          ITSN1
          Gene Name:
          intersectin 1
          Gene Aliases:
          ITSN, SH3D1A, SH3P17
          Chromosome Location:
          Chr.21: 33642400 - 33899861 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01051875_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITSN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003024.2 NP_003015.2 29 - 30 3928 102
          XM_005261026.2 XP_005261083.1 28 - 29 3991 102
          XM_017028428.1 XP_016883917.1 30 - 31 3926 102
          XM_017028429.1 XP_016883918.1 29 - 30 3895 102
          XM_017028430.1 XP_016883919.1 28 - 29 3880 102
          XM_017028431.1 XP_016883920.1 29 - 30 4006 102
          XM_017028432.1 XP_016883921.1 28 - 29 3991 102
          XM_017028433.1 XP_016883922.1 27 - 28 3793 102
          XM_017028434.1 XP_016883923.1 26 - 27 3778 102
          XM_017028435.1 XP_016883924.1 27 - 28 3682 102
          XM_017028436.1 XP_016883925.1 26 - 27 3667 102
          XM_017028437.1 XP_016883926.1 24 - 25 3346 102
          GenBank mRNA AF064244.1 - 29 - 30 3766 102
          AF114487.1 - 29 - 30 3928 102
          BC116185.1 - 28 - 29 3724 40073780 102
          BC116186.2 - 29 - 30 3739 40073781 102
          EU117382.1 - 26 - 27 3479 102
          EU120733.1 - 27 - 28 3646 102
          EU120734.1 - 27 - 28 3646 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC21000131.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ITSN1

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          regulation of Rho protein signal transduction
          positive regulation of phosphorylation
          positive regulation of apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of GTPase activity
          ephrin receptor signaling pathway
          synaptic vesicle endocytosis
          regulation of small GTPase mediated signal transduction
          positive regulation of protein kinase B signaling
          guanyl-nucleotide exchange factor activity
          calcium ion binding
          protein binding
          kinase activator activity
          molecular adaptor activity
          proline-rich region binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 TC21000131.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000381285.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000381288.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000381318.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000399326.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000399367.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 ENST00000437442.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR21001091.hg.1 NM_003024.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000381285.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000381318.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000399367.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000475422.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000489261.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000495656.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 NM_003024.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000381318.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 TC2100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000381285.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000399367.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000475422.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 NM_003024.2 Refseq
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000489261.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100146973.hg.1 ENST00000495656.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0