Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01052880_g1
          See other EFCAB8 GE Assays ›
          Gene Symbol:
          EFCAB8
          Gene Name:
          EF-hand calcium binding domain 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 32860995 - 32961845 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01052880_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EFCAB8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011529121.1 XP_011527423.1 25 - 26 3326 65

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Process(es)

          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3326115 - -
          Array Name Probe IDs Targets Target Type
          Human Genome U133 Plus 2.0 Array 227547_PM_at - -
          Human Genome U133 Plus 2.0 Array 239729_PM_at - -
          Human Genome U133 Plus 2.0 Array 239729_at - -
          Human Genome U133 Plus 2.0 Array 227547_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 227547_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239729_at8 - -
          Human Genome U95 Set 58646_at - -
          Human Genome U95 Set 58646_PM_at - -
          Human Genome U95 Set 85108_PM_at - -
          Human Genome U95 Set 85108_at - -
          Human Genome U133 Set 227547_PM_at - -
          Human Genome U133 Set 227547_at - -
          Human Genome U133 Set 239729_at - -
          Human Genome U133 Set 239729_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1667768 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1667768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0