Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001448.hg.1 |
ENST00000379341.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001448.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001448.hg.1 |
TC0X000110.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000379349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
ENST00000495599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006794.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
ENST00000439422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
TC0X00006794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
NM_006406.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
ENST00000379341.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00146501.hg.1 |
ENST00000439422.1 |
Ensembl |