Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01060528_m1
          See other PGAP2 GE Assays ›
          Gene Symbol:
          PGAP2
          Gene Name:
          post-GPI attachment to proteins 2
          Gene Aliases:
          CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
          Chromosome Location:
          Chr.11: 3797724 - 3826371 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01060528_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PGAP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256235.1 NP_001243164.1 4 - 5 298 63
          NR_027017.2 4 - 5 420 63
          NR_104271.1 4 - 5 442 63
          XM_006718183.3 XP_006718246.1 4 - 5 365 63
          XM_006718185.2 XP_006718248.1 3 - 4 257 63
          XM_006718186.2 XP_006718249.1 4 - 5 417 63
          XM_011519992.1 XP_011518294.1 5 - 6 596 63
          XM_011519995.2 XP_011518297.1 4 - 5 747 63
          XM_011519996.1 XP_011518298.1 5 - 6 548 63
          XM_011519997.1 XP_011518299.1 5 - 6 889 63
          XM_011519998.2 XP_011518300.1 5 - 6 567 63
          XM_011519999.1 XP_011518301.1 5 - 6 526 63
          XM_011520000.1 XP_011518302.1 4 - 5 440 63
          XM_011520005.2 XP_011518307.1 3 - 4 328 63
          XM_017017558.1 XP_016873047.1 3 - 4 554 63
          GenBank mRNA AK301991.1 - 4 - 5 390 63
          AL096753.1 - 4 - 5 298 63
          BI911526.1 - 4 - 5 331 63
          KU178609.1 - 2 - 3 246 63
          KU178610.1 - 2 - 3 246 63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY057920.1 - 4 - 5 345 Chr. 11 - 3797918 - 3824097 [+] on Build GRCh38 63
          GenBankmRNA BQ068222.1 - 346 Chr. - - [-] on Build 63
          GenBankmRNA HY079050.1 - 3 - 4 169 Chr. 11 - 3797917 - 3824368 [+] on Build GRCh38 63
          GenBankmRNA HY027463.1 - 4 - 5 354 Chr. 11 - 3797907 - 3824037 [+] on Build GRCh38 63
          GenBankmRNA HY050301.1 - 4 - 5 343 Chr. 11 - 3797919 - 3823923 [+] on Build GRCh38 63
          GenBankmRNA HY021670.1 - 4 - 5 333 Chr. 11 - 3797929 - 3824021 [+] on Build GRCh38 63
          GenBankmRNA HY078679.1 - 4 - 5 348 Chr. 11 - 3797913 - 3823984 [+] on Build GRCh38 63
          GenBankmRNA AA278581.1 - 1 - 2 58 Chr. 11 - 3822932 - 3824020 [+] on Build GRCh38 63
          GenBankmRNA HY086879.1 - 4 - 5 348 Chr. 11 - 3797913 - 3823947 [+] on Build GRCh38 63
          GenBankmRNA HY001256.1 - 4 - 5 354 Chr. 11 - 3797907 - 3823985 [+] on Build GRCh38 63
          GenBankmRNA BF034661.1 - 4 - 5 316 Chr. 11 - 3797945 - 3825088 [+] on Build GRCh38 63
          GenBankmRNA DC396630.1 - 4 - 5 390 Chr. 11 - 3797989 - 3824033 [+] on Build GRCh38 63
          GenBankmRNA HY005545.1 - 4 - 5 417 Chr. 11 - 3797962 - 3823964 [+] on Build GRCh38 63

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nucleoporin 98 NUP98 ADIR2,
          NUP196,
          NUP96
          -25.1kb Anti-sense Chr. 11 - 3675010 - 3797792 on NCBI Build GRCh38
          Downstream ras homolog family member G RHOG ARHG 2.8kb Anti-sense Chr. 11 - 3826978 - 3840983 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          protein transport
          signal transduction in response to DNA damage
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0