Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01064454_m1
          See other AXL GE Assays ›
          Gene Symbol:
          AXL
          Gene Name:
          AXL receptor tyrosine kinase
          Gene Aliases:
          ARK, JTK11, Tyro7, UFO
          Chromosome Location:
          Chr.19: 41219199 - 41261767 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01064454_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AXL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001699.5 NP_001690.2 9 - 10 1485 108
          GenBank mRNA M76125.1 - 9 - 10 1449 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900008143.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000576.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AXL

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          natural killer cell differentiation
          positive regulation of cytokine-mediated signaling pathway
          blood vessel remodeling
          phagocytosis
          inflammatory response
          signal transduction
          spermatogenesis
          peptidyl-tyrosine phosphorylation
          forebrain cell migration
          platelet activation
          cellular response to extracellular stimulus
          negative regulation of interferon-gamma production
          negative regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          secretion by cell
          erythrocyte homeostasis
          substrate adhesion-dependent cell spreading
          cellular response to interferon-alpha
          ovulation cycle
          apoptotic cell clearance
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          innate immune response
          viral entry into host cell
          vascular endothelial growth factor receptor signaling pathway
          cell maturation
          positive regulation of pinocytosis
          negative regulation of lymphocyte activation
          positive regulation of protein kinase B signaling
          vagina development
          cellular response to lipopolysaccharide
          dendritic cell differentiation
          negative regulation of dendritic cell apoptotic process
          virus receptor activity
          phosphatidylserine binding
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          protein binding
          ATP binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19007948.hg.1 ENST00000301178.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19007948.hg.1 ENST00000359092.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19007948.hg.1 NM_001699.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19007948.hg.1 NM_021913.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19007948.hg.1 TC19000576.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000301178.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000359092.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000593513.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000599659.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_001278599.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_001699.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_021913.4 Refseq
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 TC1900008143.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000301178.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000359092.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000593513.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 ENST00000599659.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_001278599.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_001699.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900161944.hg.1 NM_021913.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix