Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004952.hg.1 |
ENST00000296125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004952.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03004952.hg.1 |
TC03000244.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000471637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000471637.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151075.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300151075.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151075.hg.1 |
TC0300007226.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300151075.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300151075.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000296125.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000422219.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000459830.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000471637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000471637.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000490928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
ENST00000495844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007226.hg.1 |
NM_003241.3 |
Refseq |