Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024690.2 | NP_078966.2 | 74 - 75 | 42596 | 65 | |
XM_017027486.1 | XP_016882975.1 | 68 - 69 | 41676 | 65 | ||
XM_017027487.1 | XP_016882976.1 | 67 - 68 | 41610 | 65 | ||
XM_017027488.1 | XP_016882977.1 | 66 - 67 | 41589 | 65 | ||
XM_017027489.1 | XP_016882978.1 | 65 - 66 | 41522 | 65 | ||
XM_017027490.1 | XP_016882979.1 | 63 - 64 | 41211 | 65 | ||
XM_017027491.1 | XP_016882980.1 | 63 - 64 | 41211 | 65 | ||
XM_017027492.1 | XP_016882981.1 | 63 - 64 | 41208 | 65 | ||
XM_017027493.1 | XP_016882982.1 | 63 - 64 | 41208 | 65 | ||
XM_017027494.1 | XP_016882983.1 | 63 - 64 | 41208 | 65 | ||
XM_017027495.1 | XP_016882984.1 | 63 - 64 | 41208 | 65 | ||
XM_017027496.1 | XP_016882985.1 | 48 - 49 | 39807 | 65 | ||
XM_017027497.1 | XP_016882986.1 | 43 - 44 | 39333 | 65 | ||
XM_017027499.1 | XP_016882988.1 | 68 - 69 | 32463 | 65 | ||
XM_017027500.1 | XP_016882989.1 | 67 - 68 | 32382 | 65 | ||
XM_017027501.1 | XP_016882990.1 | 47 - 48 | 4390 | 65 | ||
GenBank mRNA | AF361486.3 | - | 19856 | 65 | ||
AF414442.2 | - | 65531 | 65 | |||
AK024365.1 | - | 26 - 27 | 2339 | 65 | ||
AK128856.1 | - | 30 - 31 | 2965 | 65 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |