Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000449930.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
NM_017542.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367875.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010486.hg.1 |
ENST00000367876.8 |
Ensembl |