Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL19A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001858.5 | NP_001849.2 | 48 - 49 | 3213 | 126 | |
XM_011535437.2 | XP_011533739.1 | 48 - 49 | 3480 | 126 | ||
XM_011535438.2 | XP_011533740.1 | 48 - 49 | 3208 | 126 | ||
XM_017010253.1 | XP_016865742.1 | 47 - 48 | 3438 | 126 | ||
XM_017010254.1 | XP_016865743.1 | 47 - 48 | 3426 | 126 | ||
XM_017010255.1 | XP_016865744.1 | 48 - 49 | 3419 | 126 | ||
XM_017010256.1 | XP_016865745.1 | 47 - 48 | 3171 | 126 | ||
XM_017010257.1 | XP_016865746.1 | 47 - 48 | 3354 | 126 | ||
XM_017010258.1 | XP_016865747.1 | 47 - 48 | 3087 | 126 | ||
GenBank mRNA | BC113362.1 | - | 48 - 49 | 3181 | 8322414 | 126 |
BC113364.1 | - | 48 - 49 | 3181 | 8322416 | 126 | |
D38163.1 | - | 48 - 49 | 3213 | 126 | ||
U09279.1 | - | 16 - 17 | 885 | 126 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |