Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01069063_m1
          See other SLC8A1 GE Assays ›
          Gene Symbol:
          SLC8A1
          Gene Name:
          solute carrier family 8 member A1
          Gene Aliases:
          NCX1
          Chromosome Location:
          Chr.2: 40112146 - 40512452 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01069063_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC8A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006712084.3 XP_006712147.1 2 - 3 380 98
          XM_017004750.1 XP_016860239.1 3 - 4 519 98
          XM_017004752.1 XP_016860241.1 2 - 3 320 98
          GenBank mRNA Y13032.1 - 2 - 3 337 98

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_006712084.3, XM_017004750.1, XM_017004752.1, Y13032.1 transcripts.
          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          regulation of heart rate
          regulation of sodium ion transport
          ion transport
          cellular sodium ion homeostasis
          muscle contraction
          response to nutrient
          response to xenobiotic stimulus
          response to glucose
          regulation of cell communication by electrical coupling
          positive regulation of fibroblast migration
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          vascular associated smooth muscle contraction
          telencephalon development
          positive regulation of bone mineralization
          response to ATP
          cellular response to reactive oxygen species
          sodium ion transmembrane transport
          response to immobilization stress
          response to muscle stretch
          sodium ion export across plasma membrane
          response to hydrogen peroxide
          relaxation of smooth muscle
          negative regulation of cytosolic calcium ion concentration
          cardiac muscle cell development
          calcium ion homeostasis
          relaxation of cardiac muscle
          cardiac muscle contraction
          cytosolic calcium ion transport
          calcium ion transport into cytosol
          calcium ion import
          calcium ion transmembrane transport
          cellular response to caffeine
          cellular response to cAMP
          cellular response to hypoxia
          membrane depolarization during cardiac muscle cell action potential
          cell communication by electrical coupling involved in cardiac conduction
          sodium ion import across plasma membrane
          positive regulation of the force of heart contraction
          calcium ion export
          regulation of cardiac conduction
          calcium:sodium antiporter activity
          calcium ion binding
          protein binding
          calmodulin binding
          cytoskeletal protein binding
          ankyrin binding
          transmembrane transporter binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0