Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01070332_m1
          See other IFIH1 GE Assays ›
          Gene Symbol:
          IFIH1
          Gene Name:
          interferon induced with helicase C domain 1
          Gene Aliases:
          AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1
          Chromosome Location:
          Chr.2: 162267079 - 162318708 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01070332_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IFIH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022168.3 NP_071451.2 8 - 9 2040 147
          GenBank mRNA AF095844.1 - 8 - 9 1807 147
          AK056293.1 - 2 - 3 217 147
          AK292941.1 - 9 - 10 1954 147
          AK314611.1 - 8 - 9 2035 147
          AY017378.1 - 8 - 9 1793 147
          BC078180.1 - 8 - 9 1862 147
          BC111750.1 - 8 - 9 1707 40008600 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014772.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002481.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK056293.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IFIH1

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of virus
          response to virus
          protein sumoylation
          negative regulation of type I interferon production
          positive regulation of interferon-alpha production
          positive regulation of interferon-beta production
          regulation of type III interferon production
          cytoplasmic pattern recognition receptor signaling pathway in response to virus
          MDA-5 signaling pathway
          innate immune response
          DNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          helicase activity
          protein binding
          ATP binding
          zinc ion binding
          ribonucleoprotein complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000263642.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000421365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000421365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000421365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000421365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000421365.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000464129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000464129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000464129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 ENST00000464129.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014772.hg.1 NM_022168.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0