Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
ENST00000275493.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
ENST00000395504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
ENST00000454757.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
ENST00000455089.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
ENST00000533450.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
NM_005228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07004893.hg.1 |
TC07000328.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000275493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000454757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000455089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000485503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
NM_005228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
TC0700007596.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000275493.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000454757.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000455089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
ENST00000485503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700152645.hg.1 |
NM_005228.4 |
Refseq |