Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138295.3 | NP_612152.1 | 26 - 27 | 4187 | 67 | |
XM_017011798.1 | XP_016867287.1 | 27 - 28 | 10592 | 67 | ||
GenBank mRNA | AB061683.1 | - | 26 - 27 | 4153 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |