Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01090304_m1
          See other GAS6 GE Assays ›
          Gene Symbol:
          GAS6
          Gene Name:
          growth arrest specific 6
          Gene Aliases:
          AXLLG, AXSF
          Chromosome Location:
          Chr.13: 113820549 - 113864103 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01090304_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GAS6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000820.3 NP_000811.1 13 - 14 1837 82
          GenBank mRNA AK092028.1 - 5 - 6 1809 82
          AK122969.1 - 7 - 8 1474 82
          AK126533.1 - 12 - 13 3121 82
          AK290803.1 - 13 - 14 1726 82
          AK303145.1 - 10 - 11 1383 82
          BC038984.1 - 13 - 14 1785 5761791 82
          L13720.1 - 13 - 14 1788 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300009924.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000899.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GAS6

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          positive regulation of protein phosphorylation
          positive regulation of cytokine-mediated signaling pathway
          platelet degranulation
          positive regulation of glomerular filtration
          signal peptide processing
          protein phosphorylation
          endoplasmic reticulum to Golgi vesicle-mediated transport
          phagocytosis
          apoptotic process
          cell adhesion
          signal transduction
          enzyme linked receptor protein signaling pathway
          cell population proliferation
          cellular response to starvation
          positive regulation of gene expression
          negative regulation of tumor necrosis factor-mediated signaling pathway
          macrophage cytokine production
          cell migration
          peptidyl-serine phosphorylation
          fusion of virus membrane with host plasma membrane
          viral genome replication
          platelet activation
          animal organ regeneration
          cell-substrate adhesion
          positive regulation of TOR signaling
          activation of protein kinase B activity
          negative regulation of interferon-gamma production
          negative regulation of interleukin-1 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of natural killer cell differentiation
          positive regulation of peptidyl-serine phosphorylation
          cellular response to interferon-alpha
          B cell chemotaxis
          regulation of growth
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          apoptotic cell clearance
          negative regulation of DNA-binding transcription factor activity
          protein kinase B signaling
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          viral entry into host cell
          receptor-mediated virion attachment to host cell
          positive regulation of protein export from nucleus
          positive regulation of fibroblast proliferation
          positive regulation of phagocytosis
          leukocyte migration
          positive regulation of protein kinase B signaling
          positive regulation of protein tyrosine kinase activity
          negative regulation of biomineral tissue development
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          calcium ion transmembrane transport
          cellular response to vitamin K
          cellular response to glucose stimulus
          cellular response to growth factor stimulus
          cellular response to xenobiotic stimulus
          protein localization to plasma membrane
          extracellular matrix assembly
          dendritic cell differentiation
          hematopoietic stem cell migration to bone marrow
          negative regulation of oligodendrocyte apoptotic process
          negative regulation of fibroblast apoptotic process
          negative regulation of endothelial cell apoptotic process
          positive regulation of dendritic cell chemotaxis
          negative regulation of renal albumin absorption
          negative regulation of dendritic cell apoptotic process
          phosphatidylserine binding
          signaling receptor binding
          voltage-gated calcium channel activity
          calcium ion binding
          protein binding
          protein tyrosine kinase activator activity
          receptor tyrosine kinase binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          receptor ligand activity
          molecular adaptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000355761.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000357389.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000418959.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000450766.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 ENST00000480426.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 NM_000820.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13010079.hg.1 TC13000899.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000476291.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay TC1300009924.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 NM_000820.3 Refseq
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 TC1300009924.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000327773.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000480426.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000608763.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 ENST00000610073.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300166067.hg.1 NM_000820.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix