Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015957.hg.1 |
ENST00000376365.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015957.hg.1 |
ENST00000376371.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015957.hg.1 |
NM_001135953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015957.hg.1 |
NM_025211.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015957.hg.1 |
TC09001281.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
ENST00000376365.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
ENST00000376371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
NM_001135953.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
NM_025211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
TC0900010580.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
NM_025211.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
ENST00000376365.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
ENST00000376371.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900172141.hg.1 |
NM_001135953.1 |
Refseq |