Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147205.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147205.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147205.hg.1 |
TC0200006927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200147205.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147205.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006927.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147206.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147206.hg.1 |
NR_110577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200147206.hg.1 |
TC0200006927.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200147206.hg.1 |
ENST00000433429.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200147206.hg.1 |
NR_110577.1 |
Refseq |