Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01093031_m1
          See other MUL1 GE Assays ›
          Gene Symbol:
          MUL1
          Gene Name:
          mitochondrial E3 ubiquitin protein ligase 1
          Gene Aliases:
          C1orf166, GIDE, MAPL, MULAN, RNF218
          Chromosome Location:
          Chr.1: 20499448 - 20508483 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01093031_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MUL1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024544.2 NP_078820.2 1 - 2 278 110
          GenBank mRNA AB097015.1 - 1 - 2 223 110
          AK022937.1 - 1 - 2 264 110
          AK293430.1 - 1 - 2 237 110
          AL833889.1 - 1 - 2 225 110
          BC010101.2 - 1 - 2 185 3508192 110
          BC014010.1 - 1 - 2 178 4026435 110
          EU935008.1 - 1 - 2 121 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013184.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002315.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MUL1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of mitochondrial fusion
          regulation of mitochondrion organization
          protein ubiquitination
          protein sumoylation
          negative regulation of cell growth
          protein destabilization
          positive regulation of protein sumoylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of innate immune response
          negative regulation of defense response to virus by host
          protein stabilization
          mitochondrion localization
          regulation of mitochondrial membrane potential
          negative regulation of protein kinase B signaling
          negative regulation of type I interferon-mediated signaling pathway
          cellular response to exogenous dsRNA
          negative regulation of chemokine (C-C motif) ligand 5 production
          positive regulation of mitochondrial fission
          regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          positive regulation of dendrite extension
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          ligase activity
          SUMO transferase activity
          ubiquitin protein ligase binding
          identical protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035416.hg.1 ENST00000264198.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035416.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035416.hg.1 TC01002315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01035419.hg.1 ENST00000264198.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035419.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035419.hg.1 TC01002315.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01035417.hg.1 ENST00000264198.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035417.hg.1 NM_024544.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035417.hg.1 TC01002315.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100197541.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197541.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197541.hg.1 TC0100013184.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197541.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197541.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197542.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197542.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197542.hg.1 TC0100013184.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197542.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197542.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197545.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197545.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197545.hg.1 TC0100013184.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197545.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197545.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay TC0100013184.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197543.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197543.hg.1 NM_024544.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100197543.hg.1 TC0100013184.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100197543.hg.1 ENST00000264198.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100197543.hg.1 NM_024544.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0