Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01094916_m1
          See other DST GE Assays ›
          Gene Symbol:
          DST
          Gene Name:
          dystonin
          Gene Aliases:
          BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
          Chromosome Location:
          Chr.6: 56457987 - 56954671 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01094916_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DST
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144769.2 NP_001138241.1 71 - 72 13145 62
          NM_001144770.1 NP_001138242.1 69 - 70 12816 62
          NM_015548.4 NP_056363.2 58 - 59 11700 62
          NM_183380.3 NP_899236.1 68 - 69 13090 62
          XM_005249310.3 XP_005249367.1 77 - 78 19497 62
          XM_005249315.3 XP_005249372.1 76 - 77 19444 62
          XM_005249316.2 XP_005249373.1 69 - 70 19720 62
          XM_005249318.2 XP_005249375.1 75 - 76 19158 62
          XM_005249319.2 XP_005249376.1 74 - 75 19059 62
          XM_005249320.4 XP_005249377.1 74 - 75 19115 62
          XM_005249322.4 XP_005249379.1 74 - 75 19010 62
          XM_005249323.3 XP_005249380.1 73 - 74 19343 62
          XM_005249324.2 XP_005249381.1 63 - 64 17955 62
          XM_011514824.2 XP_011513126.1 76 - 77 19408 62
          XM_011514825.2 XP_011513127.1 71 - 72 13146 62
          XM_011514826.2 XP_011513128.1 72 - 73 19016 62
          XM_017011205.1 XP_016866694.1 77 - 78 19507 62
          XM_017011206.1 XP_016866695.1 77 - 78 19507 62
          XM_017011207.1 XP_016866696.1 77 - 78 19444 62
          XM_017011208.1 XP_016866697.1 77 - 78 19507 62
          XM_017011209.1 XP_016866698.1 77 - 78 19507 62
          XM_017011210.1 XP_016866699.1 77 - 78 19507 62
          XM_017011211.1 XP_016866700.1 77 - 78 19507 62
          XM_017011212.1 XP_016866701.1 76 - 77 19273 62
          XM_017011213.1 XP_016866702.1 76 - 77 19180 62
          XM_017011214.1 XP_016866703.1 76 - 77 19180 62
          XM_017011215.1 XP_016866704.1 76 - 77 19180 62
          XM_017011216.1 XP_016866705.1 76 - 77 19180 62
          XM_017011217.1 XP_016866706.1 74 - 75 19088 62
          XM_017011218.1 XP_016866707.1 39 - 40 15116 62
          XM_017011219.1 XP_016866708.1 72 - 73 13245 62
          XM_017011220.1 XP_016866709.1 71 - 72 13188 62
          XM_017011221.1 XP_016866710.1 71 - 72 13011 62
          XM_017011222.1 XP_016866711.1 64 - 65 13404 62
          XM_017011223.1 XP_016866712.1 71 - 72 12918 62
          XM_017011224.1 XP_016866713.1 67 - 68 12758 62
          GenBank mRNA AB018271.2 - 6 - 7 990 62
          AF400226.1 - 58 - 59 11592 62
          AK096713.1 - 14 - 15 2429 62
          AK128632.1 - 17 - 18 3234 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600012123.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001828.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DST

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          cell adhesion
          integrin-mediated signaling pathway
          response to wounding
          maintenance of cell polarity
          intermediate filament cytoskeleton organization
          cell motility
          actin binding
          integrin binding
          calcium ion binding
          protein binding
          protein C-terminus binding
          microtubule plus-end binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000244364.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000312431.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000340834.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000361203.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000370754.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000370769.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000370788.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000421834.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 ENST00000446842.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 NM_015548.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06022169.hg.1 TC06001828.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000244364.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000312431.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000340834.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000361203.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000370754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000370788.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000421834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_001144769.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_001144770.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_015548.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_183380.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 TC0600012123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000370788.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000244364.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000312431.10 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000340834.8 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000361203.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000370754.9 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 ENST00000421834.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_001144769.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_001144770.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_015548.4 Refseq
          Affymetrix Human Clariom D Assay PSR0600183544.hg.1 NM_183380.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0