Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000265075.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000344624.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000382188.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000442743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000511367.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
ENST00000513349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
NM_001100412.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
NM_013235.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05017267.hg.1 |
TC05001223.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000344624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000442743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000510375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000511367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000512885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000513349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
NM_001100412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
NM_013235.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
TC0500010394.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000344624.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000442743.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000510375.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000511367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000512885.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
ENST00000513349.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
NM_001100412.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500172346.hg.1 |
NM_013235.4 |
Refseq |