Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01095067_s1
          See other LOC153546 GE Assays ›
          Gene Symbol:
          LOC153546 GOLPH3
          Gene Name:
          uncharacterized LOC153546
          golgi phosphoprotein 3
          Gene Aliases:
          -
          GOPP1, GPP34, MIDAS, Vps74
          Chromosome Location:
          -
          Chromosome Location:
          Chr.5: 32124711 - 32174319 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01095067_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC153546
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK055939.1 - 1 226 105
          Gene Symbol: GOLPH3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017009686.1 XP_016865175.1 1 105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA667279.1 - 1 226 Chr. 5 - 32173396 - 32173761 [+] on Build GRCh38 105
          GenBankmRNA DA039067.1 - 1 205 Chr. 5 - 32173413 - 32173953 [+] on Build GRCh38 105
          GenBankmRNA DB191886.1 - 1 228 Chr. 5 - 32173405 - 32173772 [+] on Build GRCh38 104
          GenBankmRNA DB004100.1 - 1 212 Chr. 5 - 32173405 - 32173660 [+] on Build GRCh38 105
          GenBankmRNA DA924707.1 - 1 222 Chr. 5 - 32173395 - 32173787 [+] on Build GRCh38 105
          GenBankmRNA DR006927.1 - 1 166 Chr. 5 - 32173452 - 32174110 [+] on Build GRCh38 105

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PDZ domain containing 2 PDZD2 AIPC,
          PAPIN,
          PDZK3,
          PIN1
          -62.5kb Sense Chr. 5 - 31639388 - 32110932 on NCBI Build GRCh38
          Downstream myotubularin related protein 12 MTMR12 3-PAP,
          PIP3AP
          51.7kb Anti-sense Chr. 5 - 32227007 - 32313008 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_017009686.1 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          Golgi organization
          cell population proliferation
          glycoprotein biosynthetic process
          protein secretion
          gene expression
          regulation of mitochondrion organization
          cell migration
          lamellipodium assembly
          positive regulation of TOR signaling
          Golgi to plasma membrane protein transport
          negative regulation of apoptotic process
          protein retention in Golgi apparatus
          Golgi vesicle budding
          positive regulation of protein secretion
          leukocyte tethering or rolling
          cell adhesion molecule production
          Golgi ribbon formation
          asymmetric Golgi ribbon formation
          protein binding
          enzyme binding
          phosphatidylinositol-4-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 721335 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 220436 BC008289.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 220436 BC014233.2 genbank_mrna_human
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3839334 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P197698 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4568 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60778 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3839334 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P197698 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P197698 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01207859 - -
          Agilent Human 1B Microarray G4111A A_32_P197698 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 236124_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500007066.hg.1 ENST00000606994.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007066.hg.1 ENST00000606994.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000265070.6 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000503610.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 ENST00000512668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Affymetrix Human Clariom D Assay TC0500010418.hg.1 NM_022130.3 Refseq
          Human Genome U133 Plus 2.0 Array 236124_PM_at - -
          Human Genome U133 Plus 2.0 Array 236124_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236124_at2 - -
          Human Genome U95 Set 46433_PM_at - -
          Human Genome U95 Set 46433_at - -
          Human Genome U133 Set 236124_at - -
          Human Genome U133 Set 236124_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229391 - -
          ABI Human Genome Survey Microarray Version 2.0 233928 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0