Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01101391_g1
          See other MAP4K4 GE Assays ›
          Gene Symbol:
          MAP4K4
          Gene Name:
          mitogen-activated protein kinase kinase kinase kinase 4
          Gene Aliases:
          FLH21957, HEL-S-31, HGK, MEKKK4, NIK
          Chromosome Location:
          Chr.2: 101697703 - 101894690 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01101391_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAP4K4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_004834.4 NP_004825.3 27 - 28 3487 142
          XM_005264050.3 XP_005264107.1 29 - 30 3702 142
          XM_005264052.3 XP_005264109.1 29 - 30 3699 142
          XM_005264057.3 XP_005264114.1 29 - 30 3633 142
          XM_005264058.3 XP_005264115.1 28 - 29 3624 142
          XM_005264059.3 XP_005264116.1 29 - 30 3540 142
          XM_005264060.3 XP_005264117.1 28 - 29 3528 142
          XM_005264062.3 XP_005264119.1 28 - 29 3471 142
          XM_005264063.3 XP_005264120.1 28 - 29 3468 142
          XM_005264064.3 XP_005264121.1 27 - 28 3462 142
          XM_005264065.3 XP_005264122.1 28 - 29 3378 142
          XM_005264066.3 XP_005264123.1 28 - 29 3375 142
          XM_005264068.3 XP_005264125.1 27 - 28 3369 142
          XM_006712865.2 XP_006712928.1 29 - 30 3609 142
          XM_017005351.1 XP_016860840.1 28 - 29 3621 142
          XM_017005356.1 XP_016860845.1 28 - 29 3531 142
          XM_017005359.1 XP_016860848.1 28 - 29 3510 142
          XM_017005360.1 XP_016860849.1 28 - 29 3507 142
          XM_017005366.1 XP_016860855.1 28 - 29 3417 142
          XM_017005369.1 XP_016860858.1 27 - 28 3276 142
          XM_017005372.1 XP_016860861.1 27 - 28 3186 142
          XM_017005373.1 XP_016860862.1 26 - 27 3177 142
          GenBank mRNA AF096300.1 - 27 - 28 3188 142

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAP4K4

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          microvillus assembly
          intracellular signal transduction
          negative regulation of apoptotic process
          regulation of JNK cascade
          creatine kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0