Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01108839_m1
          See other SEMA6C GE Assays ›
          Gene Symbol:
          SEMA6C
          Gene Name:
          semaphorin 6C
          Gene Aliases:
          SEMAY, m-SemaY, m-SemaY2
          Chromosome Location:
          Chr.1: 151131685 - 151148408 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01108839_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEMA6C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001178061.1 NP_001171532.1 6 - 7 689 78
          NM_001178062.1 NP_001171533.1 6 - 7 689 78
          NM_030913.4 NP_112175.2 6 - 7 689 78
          XM_005244835.3 XP_005244892.1 6 - 7 693 78
          XM_017000075.1 XP_016855564.1 6 - 7 1100 78
          XM_017000076.1 XP_016855565.1 6 - 7 1110 78
          XM_017000077.1 XP_016855566.1 7 - 8 2022 78
          XM_017000078.1 XP_016855567.1 5 - 6 1689 78
          XM_017000079.1 XP_016855568.1 7 - 8 2022 78
          XM_017000080.1 XP_016855569.1 5 - 6 1689 78
          XM_017000081.1 XP_016855570.1 7 - 8 2022 78
          GenBank mRNA AB022434.1 - 6 - 7 540 78
          AB058772.1 - 6 - 7 2045 78
          AF339152.1 - 6 - 7 482 78
          AF339154.1 - 6 - 7 482 78
          AK302862.1 - 6 - 7 656 78
          AK311613.1 - 5 - 6 484 78
          BC114521.1 - 5 - 6 429 40036286 78
          BC114623.1 - 5 - 6 429 40036285 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015771.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003219.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SEMA6C

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          axon guidance
          positive regulation of cell migration
          negative regulation of axon extension involved in axon guidance
          negative chemotaxis
          semaphorin-plexin signaling pathway
          semaphorin receptor binding
          chemorepellent activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 ENST00000341697.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 ENST00000368912.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 ENST00000368913.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 ENST00000368914.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 ENST00000392792.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 NM_001178061.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 NM_001178062.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 NM_030913.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01049500.hg.1 TC01003219.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000341697.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368912.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368913.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368914.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000485745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000613223.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000621728.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_001178061.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_001178062.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_030913.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 TC0100015771.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000341697.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368912.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368913.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000368914.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000485745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000613223.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 ENST00000621728.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_001178061.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_001178062.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100218274.hg.1 NM_030913.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0