Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01110781_g1
          See other PEBP1 GE Assays ›
          Gene Symbol:
          PEBP1
          Gene Name:
          phosphatidylethanolamine binding protein 1
          Gene Aliases:
          HCNP, HCNPpp, HEL-210, HEL-S-34, HEL-S-96, PBP, PEBP, PEBP-1, RKIP
          Chromosome Location:
          Chr.12: 118135884 - 118145588 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01110781_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PEBP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002567.3 NP_002558.1 1 - 2 459 72
          GenBank mRNA AK226006.1 - 1 - 2 238 72
          AK299414.1 - 1 - 2 218 72
          AK308056.1 - 1 - 2 217 72
          AK311927.1 - 1 - 2 230 72
          BC008714.2 - 1 - 2 232 2822606 72
          BC017396.1 - 1 - 2 253 4074533 72
          BC018799.1 - 1 - 2 223 72
          BC031102.1 - 1 - 2 221 5300366 72
          BF978793.1 - 1 - 2 301 72
          BP347345.1 - 1 - 2 484 72
          D16111.1 - 1 - 2 224 72
          EU794643.1 - 1 - 2 223 72
          HM131449.1 - 1 - 2 215 72
          X75252.1 - 1 - 2 243 72
          X85033.1 - 1 - 2 136 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200009071.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000926.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PEBP1

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of endopeptidase activity
          RNA binding
          serine-type endopeptidase inhibitor activity
          protein binding
          ATP binding
          phosphatidylethanolamine binding
          enzyme binding
          protein kinase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12012024.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012024.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012024.hg.1 TC12000926.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12012028.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012028.hg.1 ENST00000418769.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012028.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012028.hg.1 TC12000926.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12012031.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012031.hg.1 ENST00000418769.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012031.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012031.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012031.hg.1 TC12000926.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100016856.hg.1 ENST00000455475.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016856.hg.1 ENST00000455475.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010102.hg.1 ENST00000554856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012029.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012029.hg.1 ENST00000418769.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012029.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012029.hg.1 TC12000926.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012032.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012032.hg.1 ENST00000418769.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012032.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12012032.hg.1 NM_002567.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12012032.hg.1 TC12000926.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200167061.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167061.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167061.hg.1 TC1200009071.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200167061.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167061.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200013260.hg.1 ENST00000603980.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 ENST00000542939.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1200009071.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay TC1400010102.hg.1 ENST00000554856.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016856.hg.1 ENST00000455475.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100016856.hg.1 ENST00000455475.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000321694.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000539956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200012856.hg.1 ENST00000621133.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167064.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167064.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167064.hg.1 TC1200009071.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200167064.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167064.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167060.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167060.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167060.hg.1 TC1200009071.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200167060.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167060.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167065.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167065.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167065.hg.1 TC1200009071.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200167065.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167065.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167066.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167066.hg.1 NM_002567.3 Refseq
          Affymetrix Human Clariom D Assay PSR1200167066.hg.1 TC1200009071.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200167066.hg.1 ENST00000261313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200167066.hg.1 NM_002567.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 138550 - -
          ABI Human Genome Survey Microarray Version 2.0 215776 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix