Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033104.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033104.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033104.hg.1 |
TC01002148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033105.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033105.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033105.hg.1 |
TC01002148.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193851.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193851.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193851.hg.1 |
TC0100012674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100193851.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193851.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193852.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193852.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193852.hg.1 |
TC0100012674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100193852.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193852.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193849.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193849.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193849.hg.1 |
TC0100012674.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100193849.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193849.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
ENST00000377893.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012674.hg.1 |
NM_207370.3 |
Refseq |