Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055782.hg.1 |
ENST00000367504.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055782.hg.1 |
ENST00000367506.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055782.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055782.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01055782.hg.1 |
TC01003627.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
TC0100016685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100226566.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000367506.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000458395.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000465827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
ENST00000487028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_001202423.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016685.hg.1 |
NM_030934.4 |
Refseq |