Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004446.2 | NP_004437.2 | 10 - 11 | 1617 | 71 | |
GenBank mRNA | AK296755.1 | - | 10 - 11 | 1564 | 71 | |
AK307534.1 | - | 8 - 9 | 1191 | 71 | ||
AY493416.1 | - | 8 - 9 | 1164 | 71 | ||
BC034797.1 | - | 10 - 11 | 1413 | 71 | ||
BC058921.1 | - | 10 - 11 | 1454 | 71 | ||
BC105983.1 | - | 10 - 11 | 1415 | 71 | ||
BC126275.1 | - | 10 - 11 | 1485 | 8991991 | 71 | |
BC136465.1 | - | 10 - 11 | 1385 | 9020454 | 71 | |
CR933648.1 | - | 10 - 11 | 1456 | 71 | ||
JQ080075.1 | - | 10 - 11 | 1347 | 71 | ||
X54326.1 | - | 8 - 9 | 1189 | 71 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |