Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002134.hg.1 |
ENST00000249749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002134.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002134.hg.1 |
TC15000299.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002133.hg.1 |
ENST00000249749.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002133.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002133.hg.1 |
TC15000299.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000557876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
ENST00000559440.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500006981.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500148789.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500148789.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500148789.hg.1 |
TC1500006981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500148789.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500148789.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500148788.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500148788.hg.1 |
NM_019074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500148788.hg.1 |
TC1500006981.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500148788.hg.1 |
ENST00000249749.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500148788.hg.1 |
NM_019074.3 |
Refseq |