Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135924.1 | NP_001129396.1 | 26 - 27 | 5063 | 74 | |
XM_005249641.1 | XP_005249698.1 | 23 - 24 | 4177 | 74 | ||
XM_011515174.1 | XP_011513476.1 | 28 - 29 | 5100 | 74 | ||
XM_011515175.1 | XP_011513477.1 | 27 - 28 | 4962 | 74 | ||
XM_017011828.1 | XP_016867317.1 | 24 - 25 | 4395 | 74 | ||
XM_017011829.1 | XP_016867318.1 | 23 - 24 | 4064 | 74 | ||
GenBank mRNA | AK027618.1 | - | 15 - 16 | 2650 | 74 | |
BC144316.1 | - | 27 - 28 | 5447 | 74 | ||
BC144319.1 | - | 23 - 24 | 4216 | 9052839 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |