Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01125946_m1
          See other IL33 GE Assays ›
          Gene Symbol:
          IL33
          Gene Name:
          interleukin 33
          Gene Aliases:
          C9orf26, DVS27, IL1F11, NF-HEV, NFEHEV
          Chromosome Location:
          Chr.9: 6214591 - 6257983 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01125946_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IL33
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001314044.1 NP_001300973.1 6 - 7 587 175
          NM_001314045.1 NP_001300974.1 6 - 7 595 175
          NM_001314046.1 NP_001300975.1 6 - 7 582 175
          NM_001314047.1 NP_001300976.1 6 - 7 569 175
          NM_001314048.1 NP_001300977.1 5 - 6 474 175
          NM_033439.3 NP_254274.1 6 - 7 600 175
          XM_011518061.2 XP_011516363.1 5 - 6 535 175
          XM_017015285.1 XP_016870774.1 8 - 9 1328 175
          GenBank mRNA AB024518.1 - 5 - 6 539 175
          AY905581.1 - 5 - 6 522 175
          BC047085.1 - 6 - 7 577 5314040 175
          BQ878541.1 - 5 - 6 561 175
          CR407619.1 - 5 - 6 522 175
          HQ641439.1 - 4 - 5 411 175

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900006577.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000045.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene IL33

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of immunoglobulin production
          positive regulation of immunoglobulin production
          negative regulation of leukocyte migration
          negative regulation of T-helper 1 type immune response
          positive regulation of type 2 immune response
          transcription, DNA-templated
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of interferon-gamma production
          positive regulation of chemokine production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-5 production
          positive regulation of interleukin-6 production
          type 2 immune response
          positive regulation of macrophage activation
          positive regulation of transcription by RNA polymerase II
          positive regulation of inflammatory response
          defense response to virus
          extrinsic apoptotic signaling pathway
          cytokine activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 ENST00000381434.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 ENST00000417746.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 ENST00000456383.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09000594.hg.1 TC09000045.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000463336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000463336.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 TC0900006577.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900145029.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000381434.7 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000417746.6 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000456383.3 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000463336.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000463336.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 ENST00000611532.4 Ensembl
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199640.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_001199641.1 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq
          Affymetrix Human Clariom D Assay TC0900006577.hg.1 NM_033439.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix