Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018354.hg.1 |
ENST00000441325.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029539.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029539.hg.1 |
TC01006342.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029538.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01029538.hg.1 |
TC01006342.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100240517.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240517.hg.1 |
TC0100018354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100240517.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240518.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240518.hg.1 |
TC0100018354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100240518.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018354.hg.1 |
ENST00000441325.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018354.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240519.hg.1 |
ENST00000441325.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240519.hg.1 |
NR_002834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100240519.hg.1 |
TC0100018354.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100240519.hg.1 |
ENST00000441325.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100240519.hg.1 |
NR_002834.1 |
Refseq |