Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008377.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
NR_109875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
NR_109874.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
TC0500008377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000506485.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000508517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
ENST00000515570.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500157339.hg.1 |
NR_109875.1 |
Refseq |