Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000426969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
ENST00000456681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011651.hg.1 |
NR_015405.1 |
Refseq |