Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015177.hg.1 |
ENST00000450287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015177.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015177.hg.1 |
TC10001214.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015176.hg.1 |
ENST00000450287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015176.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015176.hg.1 |
TC10001214.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015175.hg.1 |
ENST00000450287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015175.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10015175.hg.1 |
TC10001214.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000171171.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171171.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171171.hg.1 |
TC1000010477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171171.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171171.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010477.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171172.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171172.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171172.hg.1 |
TC1000010477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171172.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171172.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171170.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171170.hg.1 |
NR_033842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000171170.hg.1 |
TC1000010477.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000171170.hg.1 |
ENST00000450287.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000171170.hg.1 |
NR_033842.1 |
Refseq |