Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01388515_m1
          See other LOC283387 GE Assays ›
          Gene Symbol:
          LOC283387
          Gene Name:
          uncharacterized LOC283387
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 57868056 - 57893819 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01388515_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC283387
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC032840.1 - 3 - 4 2972 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CTD small phosphatase 2 CTDSP2 OS4,
          PSR2,
          SCP2
          -22.9kb Sense Chr. 12 - 57819927 - 57846964 on NCBI Build GRCh38
          Downstream uncharacterized LOC100506844 LOC100506844 59.4kb Sense Chr. 12 - 57931449 - 57936164 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815210 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815130 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815193 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379597 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379642 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815129 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379585 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379679 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815025 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379652 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379658 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379647 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815141 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379617 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379670 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379609 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379588 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379606 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379633 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16563696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815120 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 118848 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 262476 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 99605 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 160734 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 18241 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 491908 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 694736 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 806440 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 48442 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 884184 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 543328 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3797403 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35636 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19557085 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19557085 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3797403 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19557085 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 143120 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02628518 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02628515 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11457821 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11459288 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P19293673 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02628523 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19557081 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02628522 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02628535 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19557085 - -
          Agilent Human 1B Microarray G4111A A_32_P802691 - -
          Agilent Human 1B Microarray G4111A A_32_P410503 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02628523 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19557085 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02628535 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P19293673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561233_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1200007905.hg.1 ENST00000619560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200007905.hg.1 ENST00000619560.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000548955.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010987.hg.1 ENST00000548955.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007905.hg.1 ENST00000619560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200007905.hg.1 ENST00000619560.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000499481.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000548955.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010987.hg.1 ENST00000548955.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1561233_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561233_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561233_at10 - -
          Human Genome U95 Set 69150_at - -
          Human Genome U95 Set 69150_PM_at - -
          Human Genome U133 Set 242386_x_at - -
          Human Genome U133 Set 242386_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 706667 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0